Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282192.1 | NP_001269121.1 | 3 | 1055 | ||
NM_001282193.1 | NP_001269122.1 | 2 | 889 | |||
NM_002937.4 | NP_002928.1 | 2 | 951 | |||
NM_194431.2 | NP_919412.1 | 2 | 503 | |||
GenBank mRNA | AK092422.1 | - | 2 | 951 | ||
BC015520.1 | - | 2 | 560 | 3905439 | ||
BG616902.1 | - | 2 | 500 | |||
BU594423.1 | - | 2 | 522 | |||
BX247985.1 | - | 2 | 741 | |||
BX248010.1 | - | 3 | 609 | |||
D37931.1 | - | 1 | 415 | |||
DR003809.1 | - | 2 | 506 |