Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AXIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003502.3 | NP_003493.1 | 11 | 3526 | ||
NM_181050.2 | NP_851393.1 | 10 | 3418 | |||
NR_134879.1 | 9 | 3321 | ||||
XM_011522682.2 | XP_011520984.1 | 11 | 3552 | |||
XM_011522683.2 | XP_011520985.1 | 10 | 3443 | |||
XM_011522684.2 | XP_011520986.1 | 11 | 3402 | |||
XM_011522686.1 | XP_011520988.1 | 10 | 2309 | |||
XM_017023743.1 | XP_016879232.1 | 11 | 3537 | |||
XM_017023744.1 | XP_016879233.1 | 10 | 3428 | |||
XM_017023745.1 | XP_016879234.1 | 10 | 2577 | |||
XM_017023746.1 | XP_016879235.1 | 10 | 3153 | |||
GenBank mRNA | AB208876.1 | - | 8 | 6207 | ||
AF009674.1 | - | 11 | 3252 | |||
BC017447.1 | - | 6 | 1728 | |||
BC035872.1 | - | 10 | 2567 | |||
BC044648.1 | - | 10 | 3190 | 5809104 | ||
KF615847.1 | - | 11 | 2497 | |||
KF615848.1 | - | 10 | 2389 | |||
KF615849.1 | - | 7 | 2013 | |||
KF615850.1 | - | 3 | 1942 | |||
KF615851.1 | - | 6 | 2345 | |||
KF615852.1 | - | 9 | 3321 | |||
KF615853.1 | - | 6 | 1395 | |||
KF615854.1 | - | 6 | 1242 |