Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEL1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244984.1 | NP_001231913.1 | 2 | 208 | ||
NM_005065.5 | NP_005056.3 | 2 | 208 | |||
XM_005267988.2 | XP_005268045.1 | 2 | 154 | |||
XM_005267989.3 | XP_005268046.1 | 2 | 211 | |||
GenBank mRNA | AB020335.1 | - | 2 | 136 | ||
AF052059.1 | - | 2 | 184 | |||
AK075511.1 | - | 2 | 187 | |||
AY358651.1 | - | 2 | 180 | |||
BP358857.1 | - | 2 | 208 |