Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005246818.1 | XP_005246875.1 | 7 | 883 | ||
XM_005246819.1 | XP_005246876.1 | 6 | 631 | |||
XM_006712720.3 | XP_006712783.1 | 7 | 670 | |||
XM_011511707.2 | XP_011510009.1 | 8 | 1011 | |||
XM_011511708.1 | XP_011510010.1 | 7 | 945 | |||
XM_011511709.2 | XP_011510011.1 | 7 | 713 | |||
GenBank mRNA | AB209866.1 | - | 5 | 7399 | ||
AF021834.1 | - | 7 | 827 | |||
AK313562.1 | - | 6 | 795 | |||
BC015514.1 | - | 6 | 797 | 3902987 | ||
J03225.1 | - | 6 | 689 |