Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: API5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142930.1 | NP_001136402.1 | 7 | 951 | ||
NM_001142931.1 | NP_001136403.1 | 6 | 789 | |||
NM_006595.3 | NP_006586.1 | 7 | 951 | |||
NR_024625.1 | 8 | 1013 | ||||
XM_006718359.3 | XP_006718422.1 | 7 | 941 | |||
XM_017018464.1 | XP_016873953.1 | 8 | 998 | |||
XM_017018465.1 | XP_016873954.1 | 8 | 998 | |||
GenBank mRNA | AF229253.1 | - | 6 | 565 | ||
AF229254.1 | - | 7 | 891 | |||
AK294724.1 | - | 6 | 746 | |||
AK299288.1 | - | 7 | 917 | |||
AK304157.1 | - | 8 | 979 | |||
AY311389.1 | - | 7 | 890 | |||
BC017709.1 | - | 7 | 871 | 4419401 | ||
BT007093.1 | - | 7 | 778 | |||
U83857.1 | - | 7 | 873 | |||
Y15906.1 | - | 7 | 910 |