Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012098.2 | NP_036230.1 | 2 | 754 | ||
XM_006717030.3 | XP_006717093.1 | 2 | 792 | |||
GenBank mRNA | AF125175.1 | - | 1 | 275 | ||
AK312876.1 | - | 2 | 673 | |||
AY358274.1 | - | 2 | 773 | |||
BC012368.1 | - | 2 | 627 | 3865062 |