Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIDEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318807.1 | NP_001305736.1 | 7 | 1753 | ||
NM_014430.2 | NP_055245.2 | 6 | 1604 | |||
XM_011536659.2 | XP_011534961.1 | 6 | 1260 | |||
XM_017021221.1 | XP_016876710.1 | 6 | 3308 | |||
GenBank mRNA | AF190901.1 | - | 6 | 1648 | ||
AF218586.1 | - | 4 | 782 | |||
AF544398.1 | - | 4 | 565 | |||
AK000387.1 | - | 4 | 559 | |||
AK122828.1 | - | 6 | 3196 | |||
BC035970.1 | - | 4 | 592 | 4716269 | ||
CB162472.1 | - | 4 | 540 |