- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: OSBPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015550.3 | NP_056365.1 | 23 | 3528 | ||
NM_145320.2 | NP_663160.1 | 21 | 3108 | |||
NM_145321.2 | NP_663161.1 | 21 | 3093 | |||
NM_145322.2 | NP_663162.1 | 20 | 3000 | |||
NR_104111.1 | 22 | 3600 | ||||
NR_104112.1 | 21 | 3507 | ||||
XM_005249698.3 | XP_005249755.1 | 23 | 3248 | |||
XM_006715681.3 | XP_006715744.1 | 22 | 3422 | |||
XM_006715682.3 | XP_006715745.1 | 22 | 3426 | |||
XM_006715683.3 | XP_006715746.1 | 21 | 3333 | |||
XM_006715684.3 | XP_006715747.1 | 22 | 3185 | |||
XM_011515258.2 | XP_011513560.1 | 23 | 3325 | |||
XM_011515259.2 | XP_011513561.1 | 24 | 3822 | |||
XM_017011948.1 | XP_016867437.1 | 25 | 3685 | |||
XM_017011949.1 | XP_016867438.1 | 23 | 3714 | |||
GenBank mRNA | AB014604.2 | - | 23 | 3465 | ||
AF392444.1 | - | 23 | 3437 | |||
AY008372.1 | - | 23 | 3358 |