Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015557.2 | NP_056372.1 | 14 | 2183 | ||
GenBank mRNA | AF425231.1 | - | 14 | 2172 | ||
AK091386.1 | - | 5 | 659 | |||
AK127046.1 | - | 14 | 2072 | |||
AL110281.1 | - | 3 | 220 | |||
AL117491.1 | - | 4 | 376 | |||
AL137693.1 | - | 3 | 199 |