Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCSK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017573.4 | NP_060043.2 | 15 | 2633 | ||
XM_005259586.1 | XP_005259643.1 | 14 | 2386 | |||
XM_011528085.2 | XP_011526387.1 | 16 | 2858 | |||
XM_011528086.2 | XP_011526388.1 | 16 | 2831 | |||
XM_011528087.2 | XP_011526389.1 | 15 | 2684 | |||
XM_011528088.2 | XP_011526390.1 | 16 | 2510 | |||
XM_011528089.2 | XP_011526391.1 | 14 | 2576 | |||
XM_011528090.2 | XP_011526392.1 | 13 | 2495 | |||
XM_011528091.2 | XP_011526393.1 | 14 | 2331 | |||
XM_011528093.2 | XP_011526395.1 | 15 | 2733 | |||
XM_011528094.1 | XP_011526396.1 | 13 | 2465 | |||
XM_011528096.1 | XP_011526398.1 | 13 | 2193 | |||
GenBank mRNA | AK057235.1 | - | 12 | 2571 | ||
AL043305.1 | - | 1 | 272 | |||
AL133566.1 | - | 3 | 955 | |||
AY358963.1 | - | 15 | 2591 | |||
BC036354.1 | - | 5 | 1333 | |||
BC142942.1 | - | 10 | 2784 |