Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017625.2 | NP_060095.2 | 6 | 713 | ||
GenBank mRNA | AB036706.1 | - | 6 | 625 | ||
AF271386.1 | - | 6 | 636 | |||
AK000029.1 | - | 6 | 662 | |||
AK222533.1 | - | 6 | 700 | |||
AY065972.1 | - | 6 | 703 | |||
AY157361.1 | - | 6 | 715 | |||
AY157362.1 | - | 6 | 707 | |||
AY358359.1 | - | 6 | 699 | |||
AY549722.1 | - | 6 | 802 | |||
AY619691.1 | - | 6 | 623 | |||
BC020664.1 | - | 6 | 713 | 4693329 | ||
CR457224.1 | - | 5 | 597 |