Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017625.2 | NP_060095.2 | 8 | 1118 | ||
GenBank mRNA | AB036706.1 | - | 8 | 1030 | ||
AK000029.1 | - | 8 | 1067 | |||
AK222533.1 | - | 8 | 1105 | |||
AY065972.1 | - | 8 | 1108 | |||
AY157361.1 | - | 8 | 1120 | |||
AY157362.1 | - | 8 | 1112 | |||
AY358359.1 | - | 8 | 1104 | |||
AY549722.1 | - | 8 | 1207 | |||
AY619691.1 | - | 8 | 1028 | |||
AY619692.1 | - | 8 | 1085 | |||
BC020664.1 | - | 8 | 1118 | 4693329 |