Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPNPEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022098.3 | NP_071381.1 | 2 | 198 | ||
GenBank mRNA | AK301635.1 | - | 2 | 197 | ||
AK310502.1 | - | 2 | 456 | |||
AK313770.1 | - | 2 | 159 | |||
AL365514.1 | - | 2 | 181 | |||
AL834310.1 | - | 2 | 148 | |||
BC001208.1 | - | 2 | 133 | 3502251 | ||
BC001681.2 | - | 2 | 165 | 2823656 | ||
BC004989.1 | - | 2 | 165 | 2823656 | ||
BX648018.1 | - | 2 | 193 | |||
CR456442.1 | - | 2 | 134 | |||
CR457396.1 | - | 2 | 97 |