Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024082.3 | NP_076987.3 | 4 | 1007 | ||
NR_033262.1 | 4 | 1348 | ||||
XM_005274747.3 | XP_005274804.1 | 4 | 1078 | |||
XM_011531197.2 | XP_011529499.1 | 4 | 1127 | |||
GenBank mRNA | AF326350.1 | - | 3 | 991 | ||
AK074574.1 | - | 4 | 1348 |