Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other THUMPD2 SIRNA Assays ›
Gene Symbol:
THUMPD2
Gene Name:
THUMP domain containing 2
Gene Aliases:
C2orf8
Chromosome Location:
Chr.2: 39736060 - 39779276 on Build GRCh38
UniGene ID:
Hs.468254
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
121944
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: THUMPD2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321468.1
NP_001308397.1
11
1554
NM_001321469.1
NP_001308398.1
10
1446
NM_001321470.1
NP_001308399.1
10
1398
NM_001321474.1
NP_001308403.1
9
1341
NM_001321475.1
NP_001308404.1
11
1544
NM_001321477.1
NP_001308406.1
11
1639
NM_001321478.1
NP_001308407.1
11
1747
NM_001321479.1
NP_001308408.1
9
1046
NM_001321480.1
NP_001308409.1
11
1737
NM_001321481.1
NP_001308410.1
11
1649
NM_025264.4
NP_079540.2
10
1456
NR_028102.1
11
1577
NR_135659.1
12
1868
NR_135660.1
12
1770
NR_135661.1
12
1858
NR_135662.1
11
1567
NR_135663.1
10
1167
NR_135664.1
12
1760
XM_011533119.2
XP_011531421.1
7
811
XM_017005050.1
XP_016860539.1
10
1408
XM_017005051.1
XP_016860540.1
11
1506
XM_017005054.1
XP_016860543.1
10
2137
GenBank mRNA
AF380577.1
-
8
873
AF380578.1
-
8
2381
AF380579.1
-
8
1547
AK057801.1
-
9
959
AK093580.1
-
11
1548
AK226034.1
-
10
1408
AK298178.1
-
10
1343
AL832115.1
-
11
1544
BC004163.2
-
10
1417
2961526
BC013299.2
-
10
1417
2961526
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA methyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
tRNA methylation
tRNA binding
tRNA (guanine) methyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-lzswf:80/10.239.160.35:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0