Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: WNT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030761.4 | NP_110388.2 | 2 | 286 | ||
XM_011541597.2 | XP_011539899.1 | 2 | 1760 | |||
XM_011541598.2 | XP_011539900.1 | 2 | 355 | |||
XM_011541599.1 | XP_011539901.1 | 2 | 271 | |||
GenBank mRNA | AB061675.1 | - | 2 | 187 | ||
AF316543.1 | - | 2 | 226 | |||
AK296058.1 | - | 2 | 266 | |||
AY009398.1 | - | 2 | 182 | |||
AY358947.1 | - | 2 | 218 | |||
BC057781.1 | - | 2 | 377 | 30330746 | ||
BT020125.1 | - | 2 | 182 |