Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NIPA2 SIRNA Assays ›
Gene Symbol:
NIPA2
Gene Name:
non imprinted in Prader-Willi/Angelman syndrome 2
Gene Aliases:
-
Chromosome Location:
Chr.15: 22838641 - 22868384 on Build GRCh38
UniGene ID:
Hs.591003
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
121954
Validated
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NIPA2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001008860.2
NP_001008860.1
7
1207
NM_001008892.2
NP_001008892.1
6
1071
NM_001008894.2
NP_001008894.1
5
1014
NM_001184888.1
NP_001171817.1
6
1162
NM_001184889.1
NP_001171818.1
10
1554
NM_030922.6
NP_112184.4
8
1329
XM_005272546.3
XP_005272603.1
8
1562
XM_005272547.4
XP_005272604.1
8
1346
XM_005272548.3
XP_005272605.1
7
1176
XM_005272550.3
XP_005272607.1
7
1014
XM_005272552.3
XP_005272609.1
6
1276
XM_005272553.4
XP_005272610.1
6
1080
XM_006720364.2
XP_006720427.1
9
1456
XM_006720365.2
XP_006720428.1
8
1301
XM_006720366.3
XP_006720429.1
7
1167
XM_006720367.1
XP_006720430.1
7
1505
XM_011543877.2
XP_011542179.1
9
1689
XM_011543878.2
XP_011542180.1
9
1856
XM_011543879.2
XP_011542181.1
9
1427
XM_011543880.2
XP_011542182.1
7
1440
XM_017022645.1
XP_016878134.1
9
1750
XM_017022646.1
XP_016878135.1
9
1517
XM_017022647.1
XP_016878136.1
9
1465
XM_017022648.1
XP_016878137.1
9
1526
XM_017022649.1
XP_016878138.1
9
1660
XM_017022650.1
XP_016878139.1
9
1623
XM_017022651.1
XP_016878140.1
10
2044
XM_017022652.1
XP_016878141.1
9
1436
XM_017022653.1
XP_016878142.1
10
1750
XM_017022654.1
XP_016878143.1
8
1274
XM_017022655.1
XP_016878144.1
7
1272
XM_017022656.1
XP_016878145.1
7
1282
XM_017022657.1
XP_016878146.1
8
1399
XM_017022658.1
XP_016878147.1
6
1120
XM_017022659.1
XP_016878148.1
8
1370
XM_017022660.1
XP_016878149.1
6
1383
XM_017022661.1
XP_016878150.1
6
1150
XM_017022662.1
XP_016878151.1
5
1219
XM_017022663.1
XP_016878152.1
5
1024
GenBank mRNA
AK096305.1
-
7
1188
AK300843.1
-
5
957
AK313584.1
-
6
1146
AY732242.1
-
10
1475
BC000957.3
-
3
500
BC011775.2
-
6
983
3640970
U90904.1
-
1
163
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001256133, NM_001256131, NM_001256130, NM_023647, NM_001256132, and others.
Matching Taqman Gene Expression Assay:
Hs00384478_m1
Gene Ontology Categories:
Function(s)
Process(es)
magnesium ion transport
magnesium ion transmembrane transport
magnesium ion transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0