Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZRANB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286568.1 | NP_001273497.1 | 14 | 2282 | ||
NM_001286569.1 | NP_001273498.1 | 15 | 2405 | |||
NM_032143.3 | NP_115519.2 | 14 | 2187 | |||
XM_005263809.1 | XP_005263866.1 | 14 | 2141 | |||
XM_006712788.1 | XP_006712851.1 | 14 | 2115 | |||
XM_011511958.2 | XP_011510260.1 | 14 | 2630 | |||
XM_011511959.2 | XP_011510261.2 | 14 | 2195 | |||
XM_011511960.1 | XP_011510262.1 | 14 | 2163 | |||
XM_011511961.2 | XP_011510263.2 | 14 | 2129 | |||
XM_011511963.1 | XP_011510265.1 | 14 | 2134 | |||
XM_011511966.2 | XP_011510268.2 | 12 | 2013 | |||
XM_017005065.1 | XP_016860554.1 | 14 | 2134 | |||
XM_017005066.1 | XP_016860555.1 | 14 | 2193 | |||
XM_017005067.1 | XP_016860556.1 | 14 | 2141 | |||
XM_017005068.1 | XP_016860557.1 | 14 | 2115 | |||
XM_017005069.1 | XP_016860558.1 | 14 | 2163 | |||
XM_017005070.1 | XP_016860559.1 | 14 | 2163 | |||
XM_017005071.1 | XP_016860560.1 | 12 | 1934 | |||
GenBank mRNA | AK095362.1 | - | 13 | 1845 | ||
AK131303.1 | - | 8 | 1331 | |||
AK304601.1 | - | 2 | 522 | |||
AL136824.1 | - | 3 | 629 | |||
AL833322.1 | - | 15 | 2286 | |||
BC089429.1 | - | 14 | 2187 | 3077975 | ||
BX647838.1 | - | 14 | 2310 |