Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTNAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322178.1 | NP_001309107.1 | 4 | 949 | ||
NM_001322179.1 | NP_001309108.1 | 4 | 949 | |||
NM_001322180.1 | NP_001309109.1 | 3 | 755 | |||
NM_001322181.1 | NP_001309110.1 | 4 | 946 | |||
NM_001322187.1 | NP_001309116.1 | 4 | 949 | |||
NM_001322188.1 | NP_001309117.1 | 4 | 949 | |||
NM_001322189.1 | NP_001309118.1 | 5 | 1208 | |||
NM_001322190.1 | NP_001309119.1 | 4 | 949 | |||
NM_001322191.1 | NP_001309120.1 | 3 | 755 | |||
NM_033401.4 | NP_207837.2 | 4 | 949 | |||
NM_138994.4 | NP_620481.2 | 4 | 487 | |||
NR_136210.1 | 4 | 949 | ||||
NR_136213.1 | 4 | 949 | ||||
NR_136216.1 | 4 | 949 | ||||
XM_011523403.2 | XP_011521705.1 | 4 | 487 | |||
XM_017023805.1 | XP_016879294.1 | 4 | 683 | |||
GenBank mRNA | AB051550.2 | - | 4 | 888 | ||
AB100092.1 | - | 4 | 503 | |||
AB100093.1 | - | 3 | 428 | |||
AB100094.1 | - | 4 | 503 | |||
AF463518.1 | - | 3 | 651 | |||
AK057327.1 | - | 4 | 473 | |||
DB050637.1 | - | 4 | 487 |