Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BIN1 SIRNA Assays ›
Gene Symbol:
BIN1
Gene Name:
bridging integrator 1
Gene Aliases:
AMPH2, AMPHL, SH3P9
Chromosome Location:
Chr.2: 127048023 - 127107400 on Build GRCh38
UniGene ID:
Hs.193163
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
122032
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BIN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320632.1
NP_001307561.1
12, 13
1443
NM_001320633.1
NP_001307562.1
13, 14
1581
NM_001320634.1
NP_001307563.1
11, 12
1164
NM_001320640.1
NP_001307569.1
14, 15
1710
NM_001320641.1
NP_001307570.1
16, 17
1857
NM_001320642.1
NP_001307571.1
17, 18
2444
NM_004305.3
NP_004296.1
13, 14
1533
NM_139343.2
NP_647593.1
17, 18
1950
NM_139344.2
NP_647594.1
16, 17
1821
NM_139345.2
NP_647595.1
14, 15
1689
NM_139346.2
NP_647596.1
14, 15
1662
NM_139347.2
NP_647597.1
14, 15
1725
NM_139348.2
NP_647598.1
13, 14
1617
NM_139349.2
NP_647599.1
13, 14
1596
NM_139350.2
NP_647600.1
12, 13
1488
NM_139351.2
NP_647601.1
11, 12
1398
XM_005263642.3
XP_005263699.1
18, 19
2068
XM_005263643.3
XP_005263700.1
17, 18
1975
XM_005263645.2
XP_005263702.1
17, 18
1459
XM_006712425.3
XP_006712488.1
16, 17
1936
XM_006712426.3
XP_006712489.1
16, 17
1936
XM_006712427.2
XP_006712490.1
16, 17
1414
XM_006712428.3
XP_006712491.1
15, 16
1828
XM_006712429.3
XP_006712492.1
15, 16
1807
XM_006712431.3
XP_006712494.1
14, 15
1738
XM_006712432.3
XP_006712495.1
14, 15
1699
XM_006712433.3
XP_006712496.1
13, 14
1609
XM_006712434.3
XP_006712497.1
12, 13
1564
XM_011510975.2
XP_011509277.1
18, 19
2701
XM_017003819.1
XP_016859308.1
18, 19
1882
XM_017003820.1
XP_016859309.1
14, 15
1798
XM_017003821.1
XP_016859310.1
13, 14
1693
XM_017003822.1
XP_016859311.1
12, 13
1600
XM_017003823.1
XP_016859312.1
13, 14
1153
XM_017003824.1
XP_016859313.1
12, 13
1084
XM_017003825.1
XP_016859314.1
12, 13
1378
XM_017003826.1
XP_016859315.1
12, 13
1045
GenBank mRNA
AF001383.1
-
13, 14
1404
AF004015.1
-
17, 18
1566
AF043898.1
-
14, 15
1341
AF043899.1
-
14, 15
1305
AF043900.1
-
13, 14
1212
AF043901.1
-
16, 17
1437
AF068914.1
-
12, 13
1164
AF068915.1
-
11, 12
1074
AF068916.1
-
12, 13
1119
AF068917.1
-
13, 14
1293
AF068918.1
-
14, 15
1338
AF070576.1
-
13, 14
1172
AK127325.1
-
3, 4
4433
AK295300.1
-
13, 14
1299
AK301153.1
-
11, 12
1164
AL713697.1
-
17, 18
1911
BC004101.2
-
12, 13
1164
3688734
BM805510.1
-
3, 4
288
BT006865.1
-
12, 13
1104
DR761993.1
-
5, 6
557
U68485.1
-
13, 14
1195
U87558.1
-
17, 18
1587
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
nucleus organization
cell proliferation
viral process
regulation of endocytosis
muscle cell differentiation
positive regulation of apoptotic process
regulation of neuron differentiation
positive regulation of astrocyte differentiation
lipid tube assembly
regulation of cell cycle arrest
protein binding
identical protein binding
tau protein binding
actin filament binding
RNA polymerase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-n6vvh:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0