Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BIN1 SIRNA Assays ›
Gene Symbol:
BIN1
Gene Name:
bridging integrator 1
Gene Aliases:
AMPH2, AMPHL, SH3P9
Chromosome Location:
Chr.2: 127048023 - 127107400 on Build GRCh38
UniGene ID:
Hs.193163
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
122033
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BIN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320632.1
NP_001307561.1
14
2085
NM_001320633.1
NP_001307562.1
15
2223
NM_001320634.1
NP_001307563.1
13
1806
NM_001320640.1
NP_001307569.1
16
2352
NM_001320641.1
NP_001307570.1
18
2499
NM_001320642.1
NP_001307571.1
19
3086
NM_004305.3
NP_004296.1
15
2175
NM_139343.2
NP_647593.1
19
2592
NM_139344.2
NP_647594.1
18
2463
NM_139345.2
NP_647595.1
16
2331
NM_139346.2
NP_647596.1
16
2304
NM_139347.2
NP_647597.1
16
2367
NM_139348.2
NP_647598.1
15
2259
NM_139349.2
NP_647599.1
15
2238
NM_139350.2
NP_647600.1
14
2130
NM_139351.2
NP_647601.1
13
2040
XM_005263642.3
XP_005263699.1
20
2710
XM_005263643.3
XP_005263700.1
19
2617
XM_005263645.2
XP_005263702.1
19
2101
XM_006712425.3
XP_006712488.1
18
2578
XM_006712426.3
XP_006712489.1
18
2578
XM_006712427.2
XP_006712490.1
18
2056
XM_006712428.3
XP_006712491.1
17
2470
XM_006712429.3
XP_006712492.1
17
2449
XM_006712431.3
XP_006712494.1
16
2380
XM_006712432.3
XP_006712495.1
16
2341
XM_006712433.3
XP_006712496.1
15
2251
XM_006712434.3
XP_006712497.1
14
2206
XM_011510975.2
XP_011509277.1
20
3343
XM_017003819.1
XP_016859308.1
20
2524
XM_017003820.1
XP_016859309.1
16
2440
XM_017003821.1
XP_016859310.1
15
2335
XM_017003822.1
XP_016859311.1
14
2242
XM_017003823.1
XP_016859312.1
15
1795
XM_017003824.1
XP_016859313.1
14
1726
XM_017003825.1
XP_016859314.1
14
2020
XM_017003826.1
XP_016859315.1
14
1687
XM_017003827.1
XP_016859316.1
13
2038
XM_017003828.1
XP_016859317.1
12
1993
GenBank mRNA
AF001383.1
-
15
2046
AF068914.1
-
14
1805
AF068915.1
-
13
1715
AF068916.1
-
14
1760
AF068917.1
-
15
1934
AF068918.1
-
16
1979
AF070576.1
-
15
1814
AK127325.1
-
5
5075
AL713697.1
-
19
2553
BC004101.2
-
14
1806
3688734
U68485.1
-
15
1836
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
nucleus organization
cell proliferation
viral process
regulation of endocytosis
muscle cell differentiation
positive regulation of apoptotic process
regulation of neuron differentiation
positive regulation of astrocyte differentiation
lipid tube assembly
regulation of cell cycle arrest
protein binding
identical protein binding
tau protein binding
actin filament binding
RNA polymerase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-k9xzd:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0