Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LONRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152271.3 | NP_689484.3 | 5 | 1228 | ||
XM_005273685.3 | XP_005273742.1 | 5 | 1230 | |||
XM_011544694.2 | XP_011542996.1 | 5 | 980 | |||
GenBank mRNA | AA284268.1 | - | 2 | 194 | ||
AK297266.1 | - | 4 | 365 | |||
BC106042.1 | - | 5 | 494 | |||
BC117381.1 | - | 2 | 159 | 40125932 | ||
BC117385.1 | - | 2 | 159 | 40125936 |