Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042439.2 | NP_001035904.1 | 1, 2 | 351 | ||
NM_178466.4 | NP_848561.2 | 1, 2 | 351 | |||
GenBank mRNA | BC066354.1 | - | 1, 2 | 309 | 4826101 | |
BF966133.1 | - | 1, 2 | 313 | |||
HY050111.1 | - | 1, 2 | 310 |