Gene Symbol: TANK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199135.1 | NP_001186064.1 | 3 | 221 | ||
NM_004180.2 | NP_004171.2 | 3 | 271 | |||
NM_133484.1 | NP_597841.1 | 3 | 271 | |||
XM_005246207.3 | XP_005246264.2 | 3 | 259 | |||
XM_017003093.1 | XP_016858582.1 | 4 | 452 | |||
XM_017003094.1 | XP_016858583.1 | 5 | 794 | |||
XM_017003095.1 | XP_016858584.1 | 5 | 699 | |||
XM_017003096.1 | XP_016858585.1 | 3 | 253 | |||
XM_017003097.1 | XP_016858586.1 | 5 | 527 | |||
GenBank mRNA | AK313096.1 | - | 3 | 221 | ||
BC003388.1 | - | 2 | 155 | |||
BC067779.1 | - | 3 | 213 | 5296558 | ||
BT009855.1 | - | 2 | 113 | |||
U59863.1 | - | 3 | 271 | |||
U63830.1 | - | 3 | 270 |