Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTNAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003632.2 | NP_003623.1 | 3, 4 | 589 | ||
XM_005257748.4 | XP_005257805.1 | 3, 4 | 1354 | |||
XM_017025238.1 | XP_016880727.1 | 5, 6 | 1724 | |||
GenBank mRNA | U87223.1 | - | 3, 4 | 590 |