Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013231.4 | NP_037363.1 | 2 | 1246 | ||
XM_005267490.3 | XP_005267547.1 | 2 | 1345 | |||
XM_011536611.2 | XP_011534913.1 | 2 | 986 | |||
XM_017021129.1 | XP_016876618.1 | 2 | 1527 | |||
XM_017021130.1 | XP_016876619.1 | 2 | 10801 | |||
XM_017021131.1 | XP_016876620.1 | 3 | 9955 | |||
XM_017021132.1 | XP_016876621.1 | 3 | 1434 | |||
XM_017021133.1 | XP_016876622.1 | 3 | 1452 | |||
XM_017021134.1 | XP_016876623.1 | 3 | 1379 | |||
XM_017021135.1 | XP_016876624.1 | 2 | 1247 | |||
GenBank mRNA | AF169676.1 | - | 1 | 536 | ||
AK056649.1 | - | 2 | 1246 | |||
AY358287.1 | - | 1 | 742 | |||
BC126249.1 | - | 2 | 1162 | 8991965 | ||
BC130290.1 | - | 2 | 1162 | 40146321 | ||
BC143936.1 | - | 2 | 1184 | 9052454 |