Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TAZ SIRNA Assays ›
Gene Symbol:
TAZ
Gene Name:
tafazzin
Gene Aliases:
BTHS, CMD3A, EFE, EFE2, G4.5, LVNCX, Taz1
Chromosome Location:
Chr.X: 154411406 - 154421726 on Build GRCh38
UniGene ID:
Hs.409911
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
122502
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TAZ
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000116.4
NP_000107.1
11
1127
NM_001303465.1
NP_001290394.1
10
1139
NM_181311.3
NP_851828.1
10
1037
NM_181312.3
NP_851829.1
10
1085
NM_181313.3
NP_851830.1
9
995
NR_024048.2
10
1143
XM_006724836.1
XP_006724899.1
11
1158
XM_006724837.1
XP_006724900.1
8
1143
XM_006724839.1
XP_006724902.1
9
1026
XM_006724841.3
XP_006724904.1
11
879
XM_006724842.3
XP_006724905.1
10
789
XM_011531189.1
XP_011529491.1
8
945
XM_011531191.2
XP_011529493.1
10
1317
XM_017029761.1
XP_016885250.1
8
1089
XM_017029762.1
XP_016885251.1
10
1068
XM_017029763.1
XP_016885252.1
8
891
XM_017029764.1
XP_016885253.1
9
656
XM_017029765.1
XP_016885254.1
9
1194
GenBank mRNA
AK130900.1
-
5
1490
AK291848.1
-
11
995
AY231461.1
-
10
719
AY231462.1
-
10
767
AY231463.1
-
9
677
AY231464.1
-
11
809
AY258036.1
-
11
809
AY258037.1
-
9
677
AY258038.1
-
10
767
AY258039.1
-
10
719
BC005062.1
-
10
1100
BC011515.1
-
7
808
4339983
CA488175.1
-
9
805
DQ884408.1
-
10
813
KU178348.1
-
7
711
KU178349.1
-
11
801
KU178350.1
-
10
759
KU178351.1
-
9
756
KU178352.1
-
10
711
KU178353.1
-
9
669
X92762.1
-
11
1089
Back To Top
More Information
Panther Classification:
Molecular Function -
acyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
heart development
skeletal muscle tissue development
hemopoiesis
cardiolipin biosynthetic process
mitochondrial respiratory chain complex I assembly
cardiolipin acyl-chain remodeling
cristae formation
mitochondrial ATP synthesis coupled electron transport
cardiac muscle tissue development
cardiac muscle contraction
1-acylglycerol-3-phosphate O-acyltransferase activity
O-acyltransferase activity
1-acylglycerophosphocholine O-acyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-ftf7j:80/10.239.160.32:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0