Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX7A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001865.3 | NP_001856.2 | 2 | 284 | ||
NR_029466.1 | 2 | 284 | ||||
GenBank mRNA | AK312154.1 | - | 2 | 139 | ||
BC100852.1 | - | 2 | 90 | 40002220 | ||
BC100853.1 | - | 2 | 90 | 40002221 | ||
BC100854.2 | - | 2 | 90 | 40002222 | ||
BC101826.1 | - | 2 | 127 | 8069332 | ||
BC101828.1 | - | 2 | 127 | 8069334 | ||
BC133654.1 | - | 2 | 90 | 40002224 | ||
CB139775.1 | - | 2 | 143 | |||
CK821341.1 | - | 2 | 295 | |||
CR407646.1 | - | 2 | 90 | |||
CR542125.1 | - | 2 | 90 | |||
HE647861.1 | - | 2 | 284 | |||
X15822.1 | - | 2 | 103 |