Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX7A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001865.3 | NP_001856.2 | 4 | 445 | ||
NR_029466.1 | 4 | 498 | ||||
GenBank mRNA | BC100852.1 | - | 4 | 251 | 40002220 | |
BC100853.1 | - | 4 | 251 | 40002221 | ||
BC100854.2 | - | 4 | 251 | 40002222 | ||
BC101826.1 | - | 4 | 288 | 8069332 | ||
BC101828.1 | - | 4 | 288 | 8069334 | ||
BC133654.1 | - | 4 | 251 | 40002224 | ||
CB139775.1 | - | 4 | 357 | |||
CK821341.1 | - | 4 | 456 | |||
HE647861.1 | - | 4 | 445 | |||
X15822.1 | - | 4 | 264 |