Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199420.3 | NP_955452.3 | 7 | 1109 | ||
XM_011512343.2 | XP_011510645.1 | 7 | 1097 | |||
XM_011512347.2 | XP_011510649.1 | 7 | 1097 | |||
XM_011512348.2 | XP_011510650.1 | 6 | 966 | |||
XM_011512352.2 | XP_011510654.1 | 7 | 1097 | |||
GenBank mRNA | AB209867.1 | - | 7 | 1037 | ||
AY032677.1 | - | 8 | 1379 | |||
CR936627.1 | - | 6 | 955 |