Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308315.1 | NP_001295244.1 | 4 | 366 | ||
NM_001308316.1 | NP_001295245.1 | 5 | 441 | |||
NM_005725.5 | NP_005716.2 | 5 | 441 | |||
XM_016999996.1 | XP_016855485.1 | 4 | 367 | |||
GenBank mRNA | AF054839.1 | - | 5 | 373 | ||
AK315385.1 | - | 5 | 401 | |||
BC021675.1 | - | 5 | 406 | 4042036 | ||
BG697236.1 | - | 1 | 23 | |||
BQ067895.1 | - | 5 | 453 | |||
CB993308.1 | - | 4 | 392 | |||
GU971730.1 | - | 5 | 373 |