Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THYN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037304.1 | NP_001032381.1 | 7 | 629 | ||
NM_001037305.1 | NP_001032382.1 | 8 | 780 | |||
NM_014174.2 | NP_054893.1 | 7 | 1162 | |||
NM_199297.1 | NP_954994.1 | 7 | 701 | |||
NM_199298.1 | NP_954995.1 | 8 | 852 | |||
XM_005271527.3 | XP_005271584.1 | 8 | 815 | |||
XM_005271528.3 | XP_005271585.1 | 7 | 664 | |||
GenBank mRNA | AF087886.1 | - | 8 | 785 | ||
AF161493.1 | - | 7 | 1124 | |||
AF182413.1 | - | 7 | 640 | |||
BC006978.1 | - | 7 | 685 | 3682324 | ||
BC093074.1 | - | 8 | 755 | 30717693 |