Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACYBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007214.1 | NP_001007215.1 | 6 | 782 | ||
NM_014412.2 | NP_055227.1 | 6 | 967 | |||
XM_017001046.1 | XP_016856535.1 | 6 | 914 | |||
GenBank mRNA | AF057356.1 | - | 6 | 656 | ||
AF314752.1 | - | 6 | 666 | |||
AK093425.1 | - | 6 | 754 | |||
AK313278.1 | - | 6 | 758 | |||
AL035305.1 | - | 6 | 659 | |||
AY423723.1 | - | 6 | 543 | |||
BC005975.1 | - | 6 | 768 | 4102572 | ||
BC022352.1 | - | 6 | 665 | 4281339 | ||
BC067823.1 | - | 6 | 649 | 5260416 | ||
BC078151.1 | - | 6 | 632 | 6065872 |