Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PGAP2 SIRNA Assays ›
Gene Symbol:
PGAP2
Gene Name:
post-GPI attachment to proteins 2
Gene Aliases:
CWH43-N, FRAG1, HPMRS3, MRT17, MRT21
Chromosome Location:
Chr.11: 3797724 - 3826371 on Build GRCh38
UniGene ID:
Hs.133968
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
122682
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PGAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145438.2
NP_001138910.1
7
918
NM_001256235.1
NP_001243164.1
8
871
NM_001256236.1
NP_001243165.1
8
1113
NM_001256237.1
NP_001243166.1
7
1009
NM_001256238.1
NP_001243167.1
7
917
NM_001256239.1
NP_001243168.1
6
848
NM_001256240.1
NP_001243169.1
6
860
NM_001283038.1
NP_001269967.1
7
930
NM_001283039.1
NP_001269968.1
7
883
NM_001283040.1
NP_001269969.1
5
502
NM_014489.3
NP_055304.1
7
1043
NR_027016.2
5
685
NR_027017.2
8
1217
NR_027018.2
8
882
NR_045923.1
6
755
NR_045925.1
6
665
NR_045926.1
6
677
NR_045927.1
6
834
NR_045929.1
6
801
NR_104270.1
6
743
NR_104271.1
8
1003
NR_104272.1
5
673
XM_006718181.3
XP_006718244.1
6
1521
XM_006718183.3
XP_006718246.1
8
938
XM_006718185.2
XP_006718248.1
7
830
XM_006718186.2
XP_006718249.1
8
990
XM_006718190.3
XP_006718253.1
7
1146
XM_006718191.3
XP_006718254.1
7
1134
XM_006718192.1
XP_006718255.1
9
1064
XM_006718193.2
XP_006718256.1
8
871
XM_011519990.2
XP_011518292.2
8
1397
XM_011519991.2
XP_011518293.2
8
1388
XM_011519992.1
XP_011518294.1
9
1169
XM_011519993.2
XP_011518295.2
8
1495
XM_011519994.2
XP_011518296.2
8
1489
XM_011519995.2
XP_011518297.1
8
1320
XM_011519996.1
XP_011518298.1
9
1121
XM_011519997.1
XP_011518299.1
9
1462
XM_011519998.2
XP_011518300.1
9
1140
XM_011519999.1
XP_011518301.1
9
1099
XM_011520000.1
XP_011518302.1
8
1013
XM_011520001.1
XP_011518303.1
7
805
XM_011520002.1
XP_011518304.1
7
793
XM_011520003.2
XP_011518305.1
8
1329
XM_011520004.2
XP_011518306.1
8
1004
XM_011520005.2
XP_011518307.1
7
901
XM_011520006.2
XP_011518308.1
8
1285
XM_011520007.2
XP_011518309.1
9
1054
XM_017017556.1
XP_016873045.1
7
884
XM_017017557.1
XP_016873046.1
7
953
XM_017017558.1
XP_016873047.1
7
1127
XM_017017559.1
XP_016873048.1
8
990
XM_017017560.1
XP_016873049.1
8
978
GenBank mRNA
AF159615.1
-
7
1045
AK292181.1
-
8
1007
AK295202.1
-
8
882
AK301991.1
-
8
1187
AK307359.1
-
6
672
AL049261.1
-
3
2072
AL096753.1
-
8
871
AL558360.3
-
6
586
AY373030.1
-
5
734
BC009930.2
-
7
805
3506614
BG468212.1
-
6
554
BI858175.1
-
2
202
BX340489.2
-
6
785
BX356273.2
-
7
837
BX448155.2
-
7
789
CD675009.1
-
4
396
CD675113.1
-
6
636
CX872317.1
-
5
613
KU178608.1
-
5
722
KU178609.1
-
6
892
KU178610.1
-
6
819
KU178611.1
-
5
734
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
GPI anchor biosynthetic process
protein transport
signal transduction in response to DNA damage
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
protein binding
protein transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-ftf7j:80/10.239.160.32:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0