Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PGAP2 SIRNA Assays ›
Gene Symbol:
PGAP2
Gene Name:
post-GPI attachment to proteins 2
Gene Aliases:
CWH43-N, FRAG1, HPMRS3, MRT17, MRT21
Chromosome Location:
Chr.11: 3797724 - 3826371 on Build GRCh38
UniGene ID:
Hs.133968
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
122683
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PGAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145438.2
NP_001138910.1
7
1205
NM_001256235.1
NP_001243164.1
8
1158
NM_001256236.1
NP_001243165.1
8
1400
NM_001256237.1
NP_001243166.1
7
1296
NM_001256238.1
NP_001243167.1
7
1204
NM_001256239.1
NP_001243168.1
6
1135
NM_001256240.1
NP_001243169.1
6
1147
NM_001283038.1
NP_001269967.1
7
1217
NM_001283039.1
NP_001269968.1
7
1170
NM_001283040.1
NP_001269969.1
5
789
NM_014489.3
NP_055304.1
7
1330
NR_027016.2
5
972
NR_027017.2
8
1504
NR_027018.2
8
1169
NR_045923.1
6
1042
NR_045925.1
6
952
NR_045926.1
6
964
NR_045927.1
6
1121
NR_045929.1
6
1088
NR_104270.1
6
1030
NR_104271.1
8
1290
NR_104272.1
5
960
XM_006718181.3
XP_006718244.1
6
1808
XM_006718183.3
XP_006718246.1
8
1225
XM_006718185.2
XP_006718248.1
7
1117
XM_006718186.2
XP_006718249.1
8
1277
XM_006718190.3
XP_006718253.1
7
1433
XM_006718191.3
XP_006718254.1
7
1421
XM_006718192.1
XP_006718255.1
9
1351
XM_006718193.2
XP_006718256.1
8
1158
XM_011519990.2
XP_011518292.2
8
1684
XM_011519991.2
XP_011518293.2
8
1675
XM_011519992.1
XP_011518294.1
9
1456
XM_011519993.2
XP_011518295.2
8
1782
XM_011519994.2
XP_011518296.2
8
1776
XM_011519995.2
XP_011518297.1
8
1607
XM_011519996.1
XP_011518298.1
9
1408
XM_011519997.1
XP_011518299.1
9
1749
XM_011519998.2
XP_011518300.1
9
1427
XM_011519999.1
XP_011518301.1
9
1386
XM_011520000.1
XP_011518302.1
8
1300
XM_011520001.1
XP_011518303.1
7
1092
XM_011520002.1
XP_011518304.1
7
1080
XM_011520003.2
XP_011518305.1
8
1616
XM_011520004.2
XP_011518306.1
8
1291
XM_011520005.2
XP_011518307.1
7
1188
XM_011520006.2
XP_011518308.1
8
1572
XM_011520007.2
XP_011518309.1
9
1341
XM_017017556.1
XP_016873045.1
7
1171
XM_017017557.1
XP_016873046.1
7
1240
XM_017017558.1
XP_016873047.1
7
1414
XM_017017559.1
XP_016873048.1
8
1277
XM_017017560.1
XP_016873049.1
8
1265
GenBank mRNA
AF159615.1
-
7
1332
AK301991.1
-
8
1474
AL096753.1
-
8
1155
BC009930.2
-
7
1092
3506614
BI858175.1
-
2
489
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
GPI anchor biosynthetic process
protein transport
signal transduction in response to DNA damage
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
protein binding
protein transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-cq5mm:80/10.239.166.46:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0