Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATG13 SIRNA Assays ›
Gene Symbol:
ATG13
Gene Name:
autophagy related 13
Gene Aliases:
KIAA0652, PARATARG8
Chromosome Location:
Chr.11: 46617276 - 46676019 on Build GRCh38
UniGene ID:
Hs.127403
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
122699
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATG13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142673.2
NP_001136145.1
7
1039
NM_001205119.1
NP_001192048.1
6
983
NM_001205120.1
NP_001192049.1
6
983
NM_001205121.1
NP_001192050.1
7
1034
NM_001205122.1
NP_001192051.1
5
901
NM_014741.4
NP_055556.2
7
1039
XM_005253262.2
XP_005253319.1
7
779
XM_005253263.2
XP_005253320.1
7
1070
XM_005253265.2
XP_005253322.1
8
1276
XM_005253266.2
XP_005253323.1
7
1005
XM_005253268.2
XP_005253325.1
5
382
XM_006718394.2
XP_006718457.1
8
1126
XM_006718395.2
XP_006718458.1
8
1061
XM_006718396.2
XP_006718459.1
7
1160
XM_011520493.1
XP_011518795.1
9
1263
XM_011520496.2
XP_011518798.1
6
1038
XM_011520499.2
XP_011518801.1
6
652
XM_011520500.1
XP_011518802.1
8
1126
XM_011520501.1
XP_011518803.1
8
1126
XM_011520502.1
XP_011518804.1
8
1126
XM_011520503.1
XP_011518805.1
8
1126
XM_017018599.1
XP_016874088.1
8
1211
XM_017018600.1
XP_016874089.1
7
779
XM_017018601.1
XP_016874090.1
6
954
XM_017018602.1
XP_016874091.1
8
1276
XM_017018603.1
XP_016874092.1
9
1393
XM_017018604.1
XP_016874093.1
7
1070
XM_017018605.1
XP_016874094.1
8
1061
XM_017018606.1
XP_016874095.1
7
1005
XM_017018607.1
XP_016874096.1
5
382
XM_017018608.1
XP_016874097.1
7
779
XM_017018609.1
XP_016874098.1
7
1070
XM_017018610.1
XP_016874099.1
6
954
XM_017018611.1
XP_016874100.1
7
1005
XM_017018612.1
XP_016874101.1
8
1276
XM_017018613.1
XP_016874102.1
7
1070
XM_017018614.1
XP_016874103.1
8
1061
XM_017018615.1
XP_016874104.1
7
1160
XM_017018616.1
XP_016874105.1
6
954
XM_017018617.1
XP_016874106.1
8
1159
XM_017018618.1
XP_016874107.1
8
1126
XM_017018619.1
XP_016874108.1
7
1010
XM_017018620.1
XP_016874109.1
7
1070
XM_017018621.1
XP_016874110.1
6
954
XM_017018622.1
XP_016874111.1
7
1022
GenBank mRNA
AB014552.1
-
6
663
AK023867.1
-
7
772
AK223603.1
-
6
659
AK289641.1
-
7
586
AK293109.1
-
7
738
AK294110.1
-
5
614
BC001331.1
-
7
468
3458359
BC002378.2
-
7
738
2961127
BC006191.2
-
7
780
3936851
CD675643.1
-
5
358
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
autophagosome assembly
macroautophagy
mitophagy in response to mitochondrial depolarization
positive regulation of protein targeting to mitochondrion
protein binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-xzndl:80/10.239.164.225:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0