Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C2CD5 SIRNA Assays ›
Gene Symbol:
C2CD5
Gene Name:
C2 calcium dependent domain containing 5
Gene Aliases:
CDP138, KIAA0528
Chromosome Location:
Chr.12: 22448546 - 22544546 on Build GRCh38
UniGene ID:
Hs.271014
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
122709
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C2CD5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286173.1
NP_001273102.1
6
817
NM_001286174.1
NP_001273103.1
5
957
NM_001286175.1
NP_001273104.1
6
817
NM_001286176.1
NP_001273105.1
6
817
NM_001286177.1
NP_001273106.1
6
817
NM_014802.2
NP_055617.1
6
817
XM_005253538.2
XP_005253595.1
6
779
XM_006719170.2
XP_006719233.1
6
779
XM_017020264.1
XP_016875753.1
6
779
XM_017020265.1
XP_016875754.1
6
779
XM_017020266.1
XP_016875755.1
6
779
XM_017020267.1
XP_016875756.1
6
779
XM_017020268.1
XP_016875757.1
6
779
XM_017020269.1
XP_016875758.1
6
779
XM_017020270.1
XP_016875759.1
6
779
XM_017020271.1
XP_016875760.1
6
779
XM_017020272.1
XP_016875761.1
6
779
XM_017020273.1
XP_016875762.1
6
779
XM_017020274.1
XP_016875763.1
6
779
XM_017020275.1
XP_016875764.1
6
779
XM_017020276.1
XP_016875765.1
6
779
XM_017020277.1
XP_016875766.1
6
779
XM_017020278.1
XP_016875767.1
6
779
GenBank mRNA
AB011100.3
-
5
570
AK295862.1
-
6
762
AK299353.1
-
5
664
AK310753.1
-
6
817
AY166851.1
-
6
789
BC042498.1
-
5
576
5174801
BC053885.1
-
1
41
BC117143.1
-
6
680
40125694
BC143859.1
-
6
680
9052375
BC143860.1
-
6
680
9052376
BC143878.1
-
6
680
9052394
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
vesicle fusion
positive regulation of glucose transport
positive regulation of vesicle fusion
cellular response to insulin stimulus
insulin receptor signaling pathway via phosphatidylinositol 3-kinase
intracellular protein transmembrane transport
protein localization to plasma membrane
positive regulation of protein targeting to membrane
positive regulation of glucose import in response to insulin stimulus
calcium ion binding
calcium-dependent phospholipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline