Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014810.4 | NP_055625.4 | 6 | 838 | ||
XM_011510191.2 | XP_011508493.1 | 6 | 797 | |||
XM_011510198.2 | XP_011508500.1 | 6 | 797 | |||
XM_017002929.1 | XP_016858418.1 | 6 | 797 | |||
XM_017002930.1 | XP_016858419.1 | 6 | 835 | |||
XM_017002931.1 | XP_016858420.1 | 6 | 797 | |||
GenBank mRNA | AF287356.1 | - | 6 | 622 | ||
AK095601.1 | - | 5 | 708 | |||
DB072323.1 | - | 3 | 315 |