Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APH1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077628.2 | NP_001071096.1 | 7 | 1348 | ||
NM_001243771.1 | NP_001230700.1 | 5 | 1515 | |||
NM_001243772.1 | NP_001230701.1 | 6 | 1138 | |||
NM_016022.3 | NP_057106.2 | 6 | 1686 | |||
NR_045033.1 | 6 | 1550 | ||||
NR_045034.1 | 4 | 1441 | ||||
NR_045035.1 | 5 | 1612 | ||||
XM_017001417.1 | XP_016856906.1 | 8 | 1760 | |||
GenBank mRNA | AF151835.1 | - | 6 | 1388 | ||
AF508787.1 | - | 6 | 1344 | |||
AK027879.1 | - | 6 | 1456 | |||
AK075295.1 | - | 6 | 1405 | |||
AK075452.1 | - | 6 | 1494 | |||
AK125685.1 | - | 1 | 1432 | |||
AK297680.1 | - | 6 | 1316 | |||
AK298832.1 | - | 6 | 945 | |||
AK298876.1 | - | 5 | 1391 | |||
AK300640.1 | - | 5 | 1320 | |||
AK307358.1 | - | 4 | 1248 | |||
AX775955.1 | - | 6 | 1422 | |||
AX775957.1 | - | 7 | 1138 | |||
AY358951.1 | - | 6 | 1490 | |||
BC001230.2 | - | 7 | 1118 | 3457593 | ||
BC008732.2 | - | 6 | 1443 | 3162330 | ||
BC009501.2 | - | 6 | 1477 | 4040230 | ||
BC015568.2 | - | 7 | 1107 | 3356401 | ||
BC017699.1 | - | 6 | 1422 | 4177580 | ||
BC020590.1 | - | 7 | 1138 | 4420942 |