Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: UTP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016037.3 | NP_057121.2 | 6 | 640 | ||
XM_011541555.2 | XP_011539857.1 | 5 | 574 | |||
GenBank mRNA | AF067802.1 | - | 6 | 603 | ||
AF151852.1 | - | 6 | 618 | |||
AK023587.1 | - | 4 | 1509 | |||
AK291900.1 | - | 6 | 570 | |||
AK308377.1 | - | 5 | 575 | |||
BC005182.1 | - | 6 | 585 | 3995984 |