Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016486.3 | NP_057570.2 | 3 | 489 | ||
GenBank mRNA | AF151063.1 | - | 3 | 468 | ||
AK024682.1 | - | 3 | 483 | |||
BC013608.1 | - | 3 | 471 | 3864518 | ||
BC034147.1 | - | 3 | 469 | 4655713 | ||
BC039718.2 | - | 2 | 392 | 5784175 | ||
BC046130.1 | - | 3 | 452 | 5787422 | ||
BC054041.1 | - | 1 | 203 | |||
BC064539.1 | - | 3 | 472 | 5725426 | ||
BC073836.1 | - | 2 | 389 | 5548608 | ||
BC121117.2 | - | 3 | 431 | 40120483 | ||
BC133692.1 | - | 3 | 430 | 40120480 |