Gene Symbol: TRNAU1AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017846.4 | NP_060316.1 | 9 | 1203 | ||
NR_003109.1 | 10 | 1351 | ||||
XM_017001542.1 | XP_016857031.1 | 8 | 1377 | |||
GenBank mRNA | BC000680.2 | - | 9 | 1127 | 3349210 |