Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018060.3 | NP_060530.3 | 1 | 262 | ||
GenBank mRNA | AK291432.1 | - | 1 | 71 | ||
AY267462.1 | - | 1 | 273 | |||
AY952883.1 | - | 1 | 175 | |||
BC137438.1 | - | 1 | 232 | 9021436 | ||
BI459732.1 | - | 1 | 266 | |||
BM149097.1 | - | 1 | 263 | |||
D28500.1 | - | 1 | 103 |