Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297760.1 | NP_001284689.1 | 5 | 1127 | ||
NM_001297761.1 | NP_001284690.1 | 4 | 867 | |||
NM_001297762.1 | NP_001284691.1 | 4 | 1004 | |||
NM_018208.3 | NP_060678.2 | 5 | 1127 | |||
XM_011509714.2 | XP_011508016.1 | 4 | 367 | |||
XM_011509715.1 | XP_011508017.1 | 4 | 672 | |||
XM_017001632.1 | XP_016857121.1 | 4 | 455 | |||
GenBank mRNA | AB073608.1 | - | 5 | 727 | ||
AK001623.1 | - | 5 | 839 | |||
AK293426.1 | - | 4 | 575 | |||
AK302145.1 | - | 4 | 604 | |||
AK316493.1 | - | 4 | 363 | |||
BC010082.2 | - | 5 | 948 | 4329734 |