Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM126B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193537.2 | NP_001180466.1 | 5, 6 | 680 | ||
NM_001193538.2 | NP_001180467.1 | 5, 6 | 721 | |||
NM_001256546.1 | NP_001243475.1 | 5, 6 | 621 | |||
NM_001256547.1 | NP_001243476.1 | 3, 4 | 427 | |||
NM_018480.4 | NP_060950.3 | 4, 5 | 549 | |||
XM_011545165.2 | XP_011543467.1 | 6, 7 | 823 | |||
GenBank mRNA | AF220193.1 | - | 5, 6 | 708 | ||
AF246240.1 | - | 4, 5 | 721 | |||
AK291150.1 | - | 5, 6 | 689 | |||
AK298189.1 | - | 4, 5 | 527 | |||
AV728059.1 | - | 3, 4 | 389 | |||
BC012065.1 | - | 5, 6 | 665 | 4575766 | ||
BC038933.1 | - | 5, 6 | 676 | 5180998 | ||
BG623309.1 | - | 3, 4 | 414 |