Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039481.1 | NP_001034570.1 | 2 | 616 | ||
NM_018638.4 | NP_061108.2 | 2 | 616 | |||
XM_005253420.4 | XP_005253477.1 | 2 | 613 | |||
XM_017019580.1 | XP_016875069.1 | 2 | 613 | |||
XM_017019581.1 | XP_016875070.1 | 2 | 613 | |||
GenBank mRNA | AF207600.2 | - | 2 | 682 | ||
AK074425.1 | - | 2 | 561 | |||
AL137750.1 | - | 2 | 565 | |||
BC006111.1 | - | 2 | 568 | |||
BC066907.1 | - | 2 | 616 | 5260109 | ||
BT007084.1 | - | 2 | 327 |