Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136204.2 | NP_001129676.1 | 12 | 3275 | ||
NM_018715.3 | NP_061185.1 | 13 | 3444 | |||
GenBank mRNA | AB040903.1 | - | 13 | 3378 | ||
AJ421269.1 | - | 13 | 3464 | |||
AK126021.1 | - | 1 | 2372 | |||
AL359612.1 | - | 10 | 2792 | |||
BC142946.1 | - | 12 | 3031 |