Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018982.4 | NP_061855.1 | 11 | 1509 | ||
NR_036639.1 | 12 | 1560 | ||||
NR_036640.1 | 11 | 1340 | ||||
NR_135075.1 | 9 | 1226 | ||||
XM_017001499.1 | XP_016856988.1 | 10 | 1267 | |||
XM_017001500.1 | XP_016856989.1 | 10 | 1330 | |||
XM_017001501.1 | XP_016856990.1 | 9 | 1111 | |||
GenBank mRNA | AK000823.1 | - | 9 | 1174 | ||
AL713668.1 | - | 11 | 1271 | |||
BC009674.1 | - | 6 | 822 | |||
BC064590.1 | - | 11 | 1475 | 5749036 |