Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195215.1 | NP_001182144.1 | 20, 21 | 1861 | ||
NR_125340.1 | 17, 18 | 1608 | ||||
XM_005244931.2 | XP_005244988.1 | 19, 20 | 1801 | |||
XM_006711192.2 | XP_006711255.1 | 21, 22 | 1771 | |||
XM_006711193.3 | XP_006711256.1 | 21, 22 | 1903 | |||
XM_006711194.3 | XP_006711257.1 | 8, 9 | 11899 | |||
XM_011509246.2 | XP_011507548.1 | 20, 21 | 1722 | |||
XM_011509247.1 | XP_011507549.1 | 19, 20 | 1662 | |||
XM_011509248.2 | XP_011507550.1 | 20, 21 | 1655 | |||
XM_011509249.2 | XP_011507551.1 | 21, 22 | 1960 | |||
XM_017000470.1 | XP_016855959.1 | 21, 22 | 1849 | |||
XM_017000471.1 | XP_016855960.1 | 20, 21 | 1711 | |||
XM_017000472.1 | XP_016855961.1 | 16, 17 | 1335 | |||
GenBank mRNA | AK000061.1 | - | 6, 7 | 398 | ||
AK091207.1 | - | 17, 18 | 1422 | |||
BC074735.2 | - | 3, 4 | 247 | |||
BP220095.1 | - | 6, 7 | 398 |