Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019083.2 | NP_061956.2 | 6 | 486 | ||
NR_135078.1 | 6 | 444 | ||||
XM_005270945.3 | XP_005271002.1 | 6 | 514 | |||
XM_005270946.2 | XP_005271003.1 | 5 | 406 | |||
XM_017001504.1 | XP_016856993.1 | 6 | 473 | |||
XM_017001505.1 | XP_016856994.1 | 6 | 462 | |||
GenBank mRNA | AK001149.1 | - | 6 | 471 | ||
AK002081.1 | - | 6 | 473 | |||
AK289404.1 | - | 6 | 473 | |||
AK294738.1 | - | 6 | 471 | |||
AK295828.1 | - | 6 | 441 | |||
AK298941.1 | - | 6 | 429 | |||
AL699548.1 | - | 5 | 417 | |||
BC057226.1 | - | 6 | 486 | |||
BC075811.1 | - | 6 | 483 | 5104751 |